Learn More
Medchemexpress LLC Milademetan (tosylate hydrate) | 2095625-97-9 | 98.9% | 808.74 | C37H44Cl2FN5O8S | 1 MG
SDP
Supplier:  Medchemexpress LLC HY101266B1MG
Encompass_Preferred
Milademetan tosylate hydrate is an orally active MDM2 inhibitor provided for preclinical and research applications. It has been studied in models of acute myeloid leukemia and solid tumors and is reported to induce G1 cell cycle arrest, senescence, and apoptosis.
  • Orally active MDM2 inhibitor mechanism suitable for pathway studies.
  • Used in preclinical research for hematologic and solid tumor models.
  • Available in solid and ready-to-use DMSO solution formats for convenience.
  • High reported purity supports consistent experimental results.
  • Characterization data (CAS, formula, molecular weight) included for traceability.
Catalog No. 50-002-12497