Learn More
Medchemexpress LLC Milademetan tosylate hydrate | 2095625-97-9 | 98.9% | 808.74 g/mol | C37H44Cl2FN5O8S | 10 MG
SDP
Supplier:  Medchemexpress LLC HY101266B10MG
Encompass_Preferred
Milademetan tosylate hydrate is the tosylate salt form of milademetan, an orally active small-molecule inhibitor of the MDM2-p53 interaction used in preclinical oncology research. Supplied as a research-use reagent, it is characterized by CAS 2095625-97-9, a molecular weight of 808.74 g/mol, and supporting documentation for laboratory use.
  • Salt form (tosylate hydrate) for improved handling and stability.
  • Orally active MDM2 inhibitor that facilitates p53 pathway reactivation.
  • High purity suitable for analytical and preclinical studies (98.9%).
  • Available in small pack sizes suitable for lab-scale experiments.
  • Includes datasheet, certificate of analysis, and safety information for lab use.
Catalog No. 50-002-12492