Learn More
Medchemexpress LLC Milademetan tosylate hydrate | 2095625-97-9 | 98.9% | 808.74 g/mol | C37H44Cl2FN5O8S | 100 MG
SDP
Supplier:  Medchemexpress LLC HY101266B100MG
Encompass_Preferred
Milademetan tosylate hydrate is the tosylate hydrate salt form of milademetan (DS-3032), an orally active MDM2 inhibitor provided as a characterized research compound for oncology studies focused on p53-MDM2 interactions. The material is supplied with analytical data and a certificate of analysis.
  • Orally active MDM2 inhibitor
  • Intended for oncology research and p53-MDM2 studies
  • High purity (98.9%) with available certificate of analysis
  • Molecular weight 808.74 g/mol
  • Supplied in measured quantities, commonly 100 MG
Catalog No. 50-002-12444