Learn More
Medchemexpress LLC Milademetan tosylate hydrate | 2095625-97-9 | 98.9% | 808.74 g/mol | C37H44Cl2FN5O8S | 25 MG
SDP
Supplier:  Medchemexpress LLC HY101266B25MG
Encompass_Preferred
Milademetan tosylate hydrate is the tosylate hydrate salt of milademetan, an orally active MDM2 inhibitor used in preclinical research. It disrupts the p53-MDM2 interaction, inducing G1 cell cycle arrest and apoptosis, and is investigated for acute myeloid leukemia and various solid tumor models. For research use only.
  • Specific MDM2 inhibitor with p53 pathway activity.
  • Induces G1 cell cycle arrest and apoptosis in preclinical models.
  • High purity reported (98.89%) on the COA.
  • Available in small research quantities, including 25 mg packaging.
  • Supplied with a datasheet and certificate of analysis for traceability.
  • Chemical identifiers: CAS 2095625-97-9; formula C37H44Cl2FN5O8S.
Catalog No. 50-002-11134