Learn More
Cayman Chemical MIlbemycIn A4 5mg
SDP
Supplier:  Cayman Chemical 171555 MG
Encompass_Preferred
The prominent member of a complex family of macrocyclic lactones produced from the fermentation of soil bacterium S. hygroscopicus subsp. aureolacrimosus; nematocide and insecticide activity via the potentiation of glutamate and GABA-gated chloride-channel opening
Catalog No. 50-296-7040