Learn More
Apexbio Technology LLC Mildronate, 50mg. Cas: 76144-81-5 MFCD: MFCD00876389. GBB hydroxylase inhibitor
SDP
Supplier:  Apexbio Technology LLC A433550
Encompass_Preferred
Mildronate is an inhibitor of biosynthesis of L-carnitine by gamma-butyrobetaine (GBB) hydroxylase and as a competitive inhibitor of renal carnitine reabsorption. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4925