Learn More
Research Products International Corp Dry Powder Milk, 1 Kilogram
SDP
Supplier:  Research Products International Corp M172001000.0
Encompass_Preferred
Nonfat dry milk for use as a blocking agent during Western Blotting to block non-specific antibody binding and reduce background signal.
Catalog No. 50-488-785