Learn More
Novus Biologicals Milk-free Antibody Diluent
SDP
Supplier:  Novus Biologicals 043524
Encompass
Milk-free Antibody Diluent for Jess, Wes, Peggy Sue and Sally Sue (20 mL). We recommend its use for dilution of goat primary, anti-goat secondary and as blocking agent as it has been shown to be superior in reducing background compared to other diluents tested. Ships at room temperature. Upon receipt, store at 2-8C.
Catalog No. NC1735422