Learn More
Grainger MILLER Handler 190 MIG Welder
Supplier:  Grainger 13Z834
Encompass_Preferred
The Hobart Handler 190 MIG Welder is a versatile, easy-to-use welder suitable for hobbyists and professionals. It can weld 24 gauge up to 1/4 inch steel and has four voltage settings to optimize your weld. The Handler 190 has a built-in spool gun connection for aluminum welding. It includes a 10 foot MIG gun, 10 foot work cable with clamp, and gas hose. This welder requires 230V input power and operates on both gas and no-gas modes. The drive system provides smooth wire feeding.
Catalog No. 50-275-1920