Learn More
Sklar Instruments Fiber Optic Miller Laryngoscope Blade, Sklar English Style, Size 00, 3mm x 40mm, Stainless Steel. EA.
SDP
Supplier:  Sklar Instruments 071462
Encompass_Preferred
Fiber Optic Miller Laryngoscope Blade, Sklar. English Style, Size 00, 3mm x 40mm, Stainless Steel. Sklar Miller Laryngoscope Blades are used for direct, rigid laryngoscopy procedures to view vocal cords, remove foreign objects from the throat, biopsy, or tracheal intubation. The Miller blade is straight and therefore longer than the curved Macintosh blade. It is inserted past the epiglottis, typically causing less trauma during intubation. The Miller blade is commonly used with pediatric and infant patients. This product is a standard, 3mm, #00 American Style Miller blade. It has a working length of 40mm and an overall length of 76mm.
Catalog No. 50-118-1383