Learn More
Sklar Instruments Miller Laryngoscope Blade, Sklar English Style, Size 4, Stainless Steel. EA.
SDP
Supplier:  Sklar Instruments 071216
Encompass_Preferred
Miller Laryngoscope Blade, Sklar. English Style, Size 4, Stainless Steel. Sklar Miller Laryngoscope Blades are used for direct, rigid laryngoscopy procedures to view vocal cords, remove foreign objects from the throat, biopsy, or tracheal intubation. The Miller blade is straight and therefore longer than the curved Macintosh blade. It is inserted past the epiglottis, typically causing less trauma during intubation. The Miller blade is commonly used with pediatric and infant patients. This product is a standard, size 4, English Style Miller blade. It has a working length of 183mm and an overall length of 205mm.
Catalog No. 50-118-1306