Learn More
GLOBAL FIA INC MILLIGAT HF PUMP W/ CER ROTOR
SDP
Supplier:  GLOBAL FIA INC MG2CERXTMD
Encompass
milliGAT HF pump with Ceramic rotor, PTFE stator, and stepper motor with integrated controller and 10x gearbox
Catalog No. NC1960934
Add to Cart
Request a Quote