Learn More
Cole-Parmer Extech 380580 Milliohm Meter, Battery-Powered
Supplier:  Cole-Parmer 2002416
Encompass
This is a high accuracy, high resolution portable Milliohm Meter with four terminal kelvin measurements and over-temperature and over-voltage protection has five ranges with 100 maximum resolution and invalid test indicators to prevent inaccurate measurements. It has also has a built-in water resistant case with a shoulder strap.
Catalog No. 50-992-375