Learn More
Apexbio Technology LLC Milnacipran HCl 101152-94-7 50mg
SDP
Supplier:  Apexbio Technology LLC B211250
Encompass_Preferred
Milnacipran hydrochloride (CAS 101152-94-7) is a small molecule classified as a serotonin-norepinephrine reuptake inhibitor (SNRI) It acts by selectively inhibiting the reuptake of both serotonin and norepinephrine in the central nervous system thereby modulating synaptic neurotransmitter levels Milnacipran HCl has been investigated for its ability to influence neurochemical pathways implicated in pain processing and mood regulation It is utilized in biomedical research to study the pathophysiology of fibromyalgia and related disorders involving monoaminergic dysfunction
Catalog No. 50-000-88732