Learn More
Cayman Chemical MIlrInon-d3 500ug
SDP
Supplier:  Cayman Chemical 25429500 UG
Encompass_Preferred
An internal standard for the quantification of milrinone by GC- or LC-MS
Catalog No. 50-293-6081