Learn More
Medchemexpress LLC Milrinone lactate | 100286-97-3 | 98.1% | C12H9N3O.xC3H6O3 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY14252
Encompass_Preferred
Milrinone lactate is a potent inotropic dilating agent that demonstrates simultaneous positive inotropic and vasodilating activities. It promotes the reduction of Systemic Vascular Resistance (SVR) and Pulmonary Vascular Resistance (PVR) in patients with Dilated Cardiomyopathy (DCM) and NYHA class III and IV heart failure. It has potential for research into cardiovascular function following cardiac surgery and in septic shock.
  • Simultaneous positive inotropic and vasodilating activities
  • Reduces SVR and PVR in heart failure patients
  • Useful for research into cardiovascular function
  • Targets phosphodiesterase (PDE)
  • For research use only
Catalog No. 50-003-54684