Learn More
Medchemexpress LLC Milvexian | 1802425-99-5 | 99.9% | 626.44 | C28H23Cl2F2N9O2 | 100MG

Supplier: Medchemexpress LLC HY125856100MG
Milvexian is an oral small-molecule factor XIa inhibitor provided for laboratory research into anticoagulation and thrombosis. It is supplied as a high-purity research-grade sample intended for in vitro and analytical studies. Not for human or clinical use.
- Oral factor XIa inhibitor for anticoagulation research.
- High purity suitable for analytical and in vitro studies.
- Molecular weight 626.44 g/mol and formula C28H23Cl2F2N9O2.
- CAS number 1802425-99-5 for unambiguous identification.
- Packaged as a 100 mg research quantity.
- Intended for research use only; not for clinical or human use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.