Learn More
MSC Milwaukee Tool I-Beam Level, 78 Inch, Magnetic, Aluminum
Supplier: MSC 14935415
This I-Beam level is designed for precise leveling applications. Constructed from durable aluminum, it features a magnetic edge for hands-free use and a length of 78 inches, suitable for various construction and calibration tasks.
- I-beam level design
- 78 inch length
- Magnetic for secure attachment
- Aluminum body material
- Red and black housing
- 3 vials for accurate readings
- 1 inch width
- 2-1/4 inch height