Learn More
Selleck Chemical LLC MINA53 inhibitor-E1064-5MG
SDP
Supplier:  Selleck Chemical LLC E10645MG
Encompass_Preferred
MINA53 inhibitor (Compound 10 2-((3-Phenylpropyl)thio)-3 4-dihydro-4-oxopyrimidine-5-carboxylic Acid) is a first-in-class inhibitor of the ribosomal oxygenase MINA53 with an IC50 of 1 5 M in vitro
Catalog No. 50-313-5306