Learn More
Crescent Chemical Co Inc Mineral Oil, 8042-47-5, MFCD00131611, 50mL
SDP
Supplier:  Crescent Chemical Co Inc SE39776.01
Encompass_Preferred
Mineral Oil, 8042-47-5, MFCD00131611, 50mL, Molecualr Biology Grade, DNase/RNase not detected. Suitable for overlaying aqueous reactions and centrifuge gradients. Storage Conditions: +15 °C TO +30 °C
Catalog No. 50-248-461