Learn More
Sigma Aldrich Fine Chemicals Biosciences Mineral oil, 8042-47-5, MFCD00131611, 100 mL
Supplier:  Sigma Aldrich Fine Chemicals Biosciences M5310100ML
Encompass
Light oil ((neat)), BioXtra, suitable for mouse embryo cell culture. Mineral oil has been commonly used and recommended for overlaying mouse embryo cultures to prevent drying during the procedure.
Catalog No. NC0921016