Learn More
BioSpec Products Mini-BeadBeater-24, 115 volt
SDP
Supplier:  BioSpec Products 112011
Encompass
The Mini-BeadBeater-24 disrupts microbial cells and plant and animal tissue by violently agitating four to twenty four 2 ml screw-cap microvials containing small glass, ceramic or steel beads and 0.1 to 1 ml disruption buffer. The performance of the Mini-BeadBeater equals or exceeds that of any other type or brand of cell disrupter. Even resistant samples like yeast, spores or fibrous tissue are completely homogenized in 1-3 minutes. The non-foaming, aerosol-free method preserves enzymes and organelles. In the presence of nucleic acid extraction media such as phenol, Gu-SCN or a commercial kit solution, DNA or RNA is recovered in the highest possible yield. The method is ideal for PAGE, PCR applications, and diagnostics using antibody or oligonucleotide probes. Because the beads and vials are disposable, there is absolutely zero cross-contamination between samples - essential for PCR techniques.
Catalog No. NC0493733