Learn More
Med Vet International Mini Blood Collection Tube - Red/Clot, J1565A, Each
Supplier:  Med Vet International JOR1565A
Encompass_Preferred
JorVet offers a wide range of blood collection tubes in various sizes and formats. All tubes adhere to the high standards of human blood collection tubes. Plastic versus Glass Human blood collection tubes have generally switched over to PET plastic tubes versus breakable glass. JorVet also uses the latest standardized color plastic caps for consistency. All tubes are in storage tray of 100 tubes Mini Tubes 1.3ml Smaller tubes for smaller patients with snap-on color coded caps and Labeled for patient info. Squared off bottom allows tube to stand upright. ?O? ring under cap for tight seal Mini Blood Collection Tube - Red/Clot
Catalog No. 50-005-81638