Learn More
Healthmark Industries Co Assure Mini Sterilization Bowie-Dick Test Pack Steam 2-1/2 X 4 Inch, 30/pk
SDP
Supplier:  Healthmark Industries Co 61301606667/DEL/DEL
Encompass
Smaller test packs that free up valuable storage and shelf space. Reduced ink and paper use during the manufacturing process. Lower shipping costs; reduced weight and smaller size of test packs. Minimizes errors and judgment calls due to unique ink technology, resulting in a complete color change. Test pack the size of a playing card and completely recyclable, post sterilization. Overall dimensions: 2.47 X 3.97 Inch.
Catalog No. NC1699851