Learn More
Supply Solutions cOmplete™, Mini, EDTA-free Protease Inhibitor Cocktail, vial of 25 tablets
Supplier:  Supply Solutions SIGMA 11836170001
Encompass
Protease Inhibitor Cocktail Tablets provided in a glass vial. cOmplete, Mini, EDTA-free Protease Inhibitor Tablets, inhibit a broad spectrum of serine and cysteine proteases. In contrast to other cOmplete tablets, they do not contain EDTA, leaving the stability and the function of metal-dependant proteins unaffected. The affinity purification of Poly-His-tagged fusion proteins using IMAC (Immobilized Metal Affinity Chromatography) is also facilitated (no dialysis necessary).
Catalog No. 50-161-3323