Learn More
System Biosciences LLC SmartSEC Mini EV Isolation System, 10 Reactions
SDP
Supplier:  System Biosciences LLC SSEC100A1
Encompass
Leverage powerful SmartSEC technology in a spin column format-isolate EVs from small volumes of biofluids from serum to Developmental Biology samples.
Catalog No. NC1853910