Learn More
Labelmaster Inc GHS Pictogram Mini Label Assortment, 25/pk
SDP
Supplier:  Labelmaster Inc GHISMLC
Encompass_Preferred
Properly label your hazardous chemicals with Labelmaster's Mini GHS Pictogram Labels. The UN Globally Harmonized System (GHS) of classification and labeling of chemicals is used to: enhance the protection of human health and the environment by providing an international system for hazard communication; provide a recognized framework for countries without a system; reduce the need for testing and evaluation of chemicals; and facilitate international trade in chemicals whose hazards have been properly assessed and identified on an international basis.
Catalog No. NC0565637