Learn More
Research Products International Corp Mini-Pipettor, 25ul, Gray with Purple Plunger
SDP
Supplier:  Research Products International Corp 347023
Encompass
New Ergonomic compact design features optimum size to facilitate ease of handling. Engineered for precise air displacement pipetting with two stage plunger and allows last drop dispensing. Available in a full range of volumes (5µl to 1000µl) and color plunger options. Unique tip cone accepts both micro tips (200µl) and ultra micro tip (up to 25µl). Built from sturdy molded polypropylene, will handle thousands of pipetting and is fully autoclavable.
Catalog No. 50-197-8539