Learn More
Epoch Life Science Inc EconoSpin® RNA Mini Spin Columns w/ lid (250 columns with collection tubes)
Supplier: Epoch Life Science Inc 1940250
Silica membrane mini spin columns with lid and collection tubes for RNA purification. Sample volume: 850 ul. Binding capacity: 45 ug RNA. Minimum elution volume: 20 ul. Compatible with third party buffer sets. 18 month shelf life. Used by labs around the world.