Learn More
Bio Rad Laboratories MINI TRANSBLOT TRANSFER CELL
Supplier:  Bio Rad Laboratories 1703930
Encompass
Mini Trans-Blot Electrophoretic Transfer Cell, Includes 2 gel holder cassettes, 4 fiber pads, modular electrode assembly, Bio-ice cooling unit, tank, lid with cables, and instructions
Catalog No. NC9411632