Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences EMS Swiss Line Mini Tweezer Style 3M, 70 mm
SDP

Supplier:  Electron Microscopy Sciences 78003M

Encompass_Preferred

Mini tweezers with edges on the outside rounded off. 70 mm long. M = 8 mm, N = 1.5 mm, P = 42 mm.
OAL: 70 mm
Width: M: 8 mm
Gap: N:1.5 mm
Tip length: P: 42 mm

Catalog No. 50-192-8658


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.