Learn More
Research Products International Corp Mini UV Viewing Cabinet, 9 x 10 x 7 Inches
SDP
Supplier:  Research Products International Corp 95007201
Encompass
Compact and portable size allows convenient use in crowded spaces. Designed for use with 6 watt Handheld UV Lamps, available separately. Cabinet features a removable bottom panel, contrast control UV filter, soft rubber viewing port and access curtain that restricts ambient light from entering cabinet interior. Constructed with a metal base and plastic top.
Catalog No. 50-998-220