Learn More
BURKLE INC Mini ViscoSampler, V4A, 39.37 in. (100 cm), volume50 ml, Ø 0.59 in. (15 mm)
Supplier:  BURKLE INC 53411100
Encompass_Preferred
Sampler for viscous substances such as pastes, mud, creams, oils. The sample is drawn in using the pump rod and directly conveyed to a sampling container. Easy cleaning for flawless sampling.
Catalog No. NC2187700