Learn More
USA Scientific Inc Mini-100 Orbital-Genie
Supplier:  USA Scientific Inc 74061000
Encompass
Mini-100 Orbital-Genie, with 11.4" x 11.4" non-slip, rubber coated platform. 20-100 rpm, 120V.
Catalog No. 50-274-4955