Learn More
World Precision Instrument Miniature Micropositioner - Three Axes-MM3
SDP
Supplier:  World Precision Instrument MM3
Encompass
The MM1 single stage measures only 5 x 11 x 26 mm with 3 mm travel. It provides precise and smooth motion with no backlash, positive spring loaded carriage, straight within 1 micron and less than 1 micron maximum wobble. It features fine 80 TPI screw adjustment. The 10 mm square mounting surface has a 3.9 mm tapped center hole for transmission and/or mounting. X-Y-Z axis configuration. Available in single X (MM1), X-Y, X-Y-Z (MM1-3) axis configurations. The unit may be purchased with an optional magnetic base (MM1-ALL or MM3-ALL.
Catalog No. 50-253-8689