Learn More
Cytiva (Formerly Pall Lab) Minimate™ EVO Tangential Flow Filtration (TFF) System
Supplier:  Cytiva OAPMPUNV
Encompass
Minimate™ TFF System Streamlines Laboratory-Scale Concentration, Desalting, and Buffer Exchange Processes- Multiple Applications from protein diafiltration, antibody recovery to viral concentration-Plug in a Minimate TFF capsule, add sample, and turn on the pump to start processing.-The use of a conical bottom reservoir and compact design enables high concentration factors from up to 1 L or more of sample to be achieved. Concentrate your sample down to as little as 5 mL.-All wetted components are made from low protein binding, chemically resistant, biologically safe materials.-Very low volume and interior fluid contact area.-Roller head peristaltic pumps provide gentle processing and are the choice for critical applications such as fragile biomolecules.-System components are designed for easy assembly and disassembly; no tools required. Tubing fitting is simple and quick. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-197-9554
Add to Cart
Request a Quote