Learn More
Bioworld MINISLOT 1010-CHANNELS)
Supplier:  Bioworld 209300091
Encompass_Preferred
9.0 x 5.5 blot dimensions. Reliably perform up to 1350 simultaneous hybridizations and antibody screening Economical for large and small scale antibody screening Requires only 50 microliter sample per channel Perform multiple screenings...
Catalog No. 50-198-530