Learn More
Medchemexpress LLC Minocycline | 10118-90-8 | MFCD00072105 | 98.6% | 457.48 | 100 MG
Supplier: Medchemexpress LLC HY17412A100MG
Minocycline is an orally active, potent, and BBB-penetrated semi-synthetic tetracycline antibiotic. It is a hypoxia-inducible factor (HIF)-1α inhibitor. Minocycline exhibits anti-cancer, anti-inflammatory, and glutamate antagonist effects. It reduces glutamate neurotransmission, demonstrating neuroprotective properties and antidepressant effects. Additionally, minocycline inhibits bacterial protein synthesis by binding with the 30S subunit of the bacterial ribosome, which leads to a bacteriostatic effect.
- Orally active
- Potent
- BBB-penetrated
- Semi-synthetic tetracycline antibiotic
- Hypoxia-inducible factor (HIF)-1α inhibitor
- Anti-cancer effects
- Anti-inflammatory effects
- Glutamate antagonist effects
- Reduces glutamate neurotransmission
- Neuroprotective properties
- Antidepressant effects
- Inhibits bacterial protein synthesis
- Bacteriostatic effect