Learn More
Medchemexpress LLC Minocycline | 10118-90-8 | MFCD00083669 | 98.6% | 10 MG
SDP
Supplier:  Medchemexpress LLC HY17412A10MG
Encompass_Preferred
Minocycline is a semi-synthetic tetracycline antibiotic provided for research and analytical applications. It is supplied in a small research pack (10 mg) with high reported purity and characterized molecular properties for use in biological screening and method development.
  • High purity (98.6%) suitable for analytical and biological assays.
  • Small research pack (10 mg) for screening and method development.
  • Orally active tetracycline that penetrates the blood-brain barrier.
  • Applicable to studies of anti-inflammatory, neuroprotective, and glutamate antagonist effects.
  • Molecular weight 457.5 g/mol; formula C23H27N3O7.
Catalog No. 50-002-98401