Learn More
Selleck Chemical LLC Minocycline HCl S4226-200mg
SDP
Supplier:  Selleck Chemical LLC S4226200MG
Encompass_Preferred
Minocycline HCl is the most lipid soluble and most active tetracycline antibiotic binds to the 30S ribosomal subunit preventing the binding of tRNA to the mRNA-ribosome complex and interfering with protein synthesis
Catalog No. 50-310-9392