Learn More
Apexbio Technology LLC Minocycline HCl(Synonyms: Minocycline hydrochloride, Minocin, Dynacin, Solodyn, Minomycin, Akamin, Arestin, Minostad), 50mg, CAS: 13614-98-7.
SDP
Supplier:  Apexbio Technology LLC B179150
Encompass_Preferred
Minocycline HCl (CAS 13614-98-7) is a semisynthetic tetracycline derivative known for its broad-spectrum antimicrobial properties and biological activity beyond antibacterial effects It acts primarily by inhibiting bacterial protein synthesis through reversible binding to the 30S ribosomal subunit thus preventing aminoacyl-tRNA from attaching to the ribosome-mRNA complex Independent of its antibiotic activity minocycline exhibits anti-inflammatory neuroprotective and antiapoptotic effects potentially mediated by suppression of cellular inflammatory pathways reduction of microglial activation and modulation of apoptotic signaling These properties make minocycline a compound of considerable interest in preclinical research for studying inflammation-related pathologies and neurodegenerative disorders
Catalog No. 50-000-60564