Learn More
Medchemexpress LLC Minocycline (hydrochloride) | 13614-98-7 | 99.1% | 493.94 | 1 ML
Supplier: Medchemexpress LLC HY1741210MM/1ML
Minocycline hydrochloride is an orally active, potent, and BBB-penetrated semi-synthetic tetracycline antibiotic. It functions as a hypoxia-inducible factor (HIF)-1α inhibitor and inhibits bacterial protein synthesis by binding to the 30S subunit of the bacterial ribosome, leading to a bacteriostatic effect. This compound also exhibits anti-cancer, anti-inflammatory, and glutamate antagonist properties.
- Orally active.
- Potent.
- Blood-brain barrier (BBB)-penetrated.
- Functions as a hypoxia-inducible factor (HIF)-1α inhibitor.
- Inhibits bacterial protein synthesis.
- Exhibits bacteriostatic effect.
- Shows anti-cancer effects.
- Demonstrates anti-inflammatory properties.
- Acts as a glutamate antagonist.
- Reduces glutamate neurotransmission.
- Possesses neuroprotective properties.
- Displays antidepressant effects.