Learn More
Medchemexpress LLC Minodronic acid | 180064-38-4 | 99.67% | 322.15 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY163225MG
Encompass_Preferred
Minodronic acid is an FPP synthase inhibitor with an IC50 of 3 nM and an antagonist of P2X2/3 receptors with an IC50 of 62.7 μM. It induces tumor cell apoptosis, inhibits cell growth, and suppresses bone resorption. It can be used in research related to osteoporosis and cancer, inhibiting proliferation in human bladder cancer cell lines and osteoclast-mediated bone resorption.
  • Inhibits FPP synthase
  • Antagonizes P2X2/3 receptors
  • Induces tumor cell apoptosis
  • Inhibits cell growth
  • Suppresses bone resorption
  • White to off-white solid appearance
Catalog No. 50-003-94121