Learn More
Medchemexpress LLC Minodronic acid hydrate | 155648-60-5 | 99.67% | 340.16 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY16322A100MG
Encompass_Preferred
Minodronic acid hydrate is a third-generation bisphosphonate that directly and indirectly prevents proliferation, induces apoptosis, and inhibits metastasis of various types of cancer cells. It also acts as an antagonist of purinergic P2X2/3 receptors, which are involved in pain.
  • Directly prevents cell proliferation.
  • Induces apoptosis in cancer cells.
  • Inhibits metastasis of various cancer cell types.
  • Antagonizes purinergic P2X2/3 receptors.
  • Involved in pain management.
Catalog No. 50-003-94122