Learn More
Cayman Chemical MInoxIdIl Sulfate 10mg
SDP
Supplier:  Cayman Chemical 3608410 MG
Encompass_Preferred
A hair follicle stimulator and an active metabolite of minoxidil; binds to KATP channels in a biphasic manner in COS-7 cells expressing the human channel (IC50s = 0.62 and 0.96 µM); induces vasodilation in isolated rabbit mesenteric artery at 5 µM
Catalog No. 50-291-1968