Learn More
Enzo Life Sciences Minoxidil-sulfate (25mg). CAS: 83701-22-8
SDP
Supplier:  Enzo Life Sciences BMLKC1300025
Encompass_Preferred
A selective ATP-sensitive K+ (KATP) channel opener. It is the active metabolite of minoxidil (BML-KC125) and is a potent (IC50=0.14 µM) vascular smooth muscle relaxant. Antihypertensive. Stimulates renin secretion. Alternative name: U-58838. Purity: ≥98%. Solubility: Soluble in DMSO (25mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-1063