Learn More
Medchemexpress LLC Minoxidil sulfate | 83701-22-8 | 98.0% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB144550MG
Encompass_Preferred
Minoxidil sulfate is a potent, ATP-sensitive K+ channel opener and the sulfated metabolite of minoxidil. It is recognized as a vasodilator that promotes hair growth in vivo. It is for research use only and not sold to patients.
  • Potent and ATP-sensitive K+ channel opener.
  • Sulfated metabolite of minoxidil.
  • Considered as a vasodilator to promote hair growth in vivo.
Catalog No. 50-004-11194