Learn More
Apexbio Technology LLC Minoxidil sulphate 83701-22-8 5g
SDP
Supplier:  Apexbio Technology LLC C65135000
Encompass_Preferred
Minoxidil sulphate (CAS 83701-22-8) is a potassium channel opener specifically acting on ATP-sensitive potassium channels It is the biologically active metabolite resulting from the sulfation of minoxidil Pharmacologically minoxidil sulphate exhibits vasodilatory activity and animal studies demonstrate its capacity to stimulate hair follicle growth Consequently it serves as an important compound in biomedical research focused on conditions involving hair regrowth and restoration
Catalog No. 50-000-63448