Learn More
MSC Vermont Gage Plug Gage, Class ZZ, Steel, 0.3340 in. Diameter, 2 in. Length, 1 ea
Supplier:  MSC 68203249
Encompass
This Class ZZ minus plug and pin gage is a precision tool designed for inspecting, calibrating, and checking dimensions of measuring devices such as micrometers and verniers. It features a 0.3340 in. diameter and an overall length of 2 in., constructed from durable steel with a black oxide finish.
  • Class ZZ accuracy with a tolerance of 0.0002 in.
  • Hardness rating of 60 to 65 Rc
  • Surface finish of 10 microinch
  • Marked with size and direction of tolerance
  • Includes certificate of accuracy with NIST traceability
Catalog No. 50-005-11937