Learn More
Invent Biotechnologies Inc Minute™ Cytoplasmic & Nuclear Extraction Kits for Cells - Spin Column-Based < 15 min (50 Preps)
Supplier:  Invent Biotechnologies Inc SC003
Encompass
Minute™ Cytoplasmic and Nuclear Extraction Kit for Cells is composed of optimized cytoplasmic extraction buffer, nuclear protein extraction buffer and protein extraction filter cartridges with 2.0 ml collection tubes. The kit is designed to quickly separate native cytosol and nuclear proteins from mammalian cells and protoplasts of plant, bacteria, yeast and fungus. Separation of cytoplasmic and nuclear proteins can be accomplished in less than 15 minutes using the protein extraction filter cartridges.
Catalog No. NC1518044